Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129227.4 | NP_001122699.2 | 1 - 2 | 44 | 143 | |
NM_001267920.3 | NP_001254849.1 | 1 - 2 | 44 | 143 | ||
NM_065077.6 | NP_497478.2 | 1 - 2 | 53 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |