Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381759.2 | NP_001367968.1 | 14 - 15 | 3104 | 80 | |
NM_001384037.1 | 18 - 19 | 4113 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |