Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065104.7 | NP_497505.3 | 6 - 7 | 1052 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |