Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065133.3 | NP_497534.1 | 1 - 2 | 210 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |