Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phf-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027738.2 | NP_001022909.1 | 2 - 3 | 170 | 79 | |
GenBank mRNA | AF537097.1 | - | 2 - 3 | 163 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |