Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027461.4 | NP_001022632.1 | 13 - 14 | 2609 | 100 | |
GenBank mRNA | AY099352.1 | - | 13 - 14 | 2611 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Caenorhabditis elegans Probe spans exons |