Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027461.4 | NP_001022632.1 | 7 - 8 | 1261 | 82 | |
NM_065139.8 | NP_497540.3 | 7 - 8 | 1261 | 82 | ||
GenBank mRNA | AY099352.1 | - | 7 - 8 | 1263 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |