Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381782.1 | NP_001367936.1 | 3 - 4 | 647 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |