Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129270.3 | NP_001122742.2 | 11 - 12 | 1474 | 55 | |
NM_065219.5 | NP_497620.3 | 3 - 4 | 317 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |