Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065250.3 | NP_497651.2 | 6 - 7 | 628 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |