Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065262.6 | NP_497663.1 | 3 - 4 | 721 | 87 | |
GenBank mRNA | AB017190.1 | - | 3 - 4 | 663 | 87 | |
U65429.1 | - | 3 - 4 | 702 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |