Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267939.3 | NP_001254868.1 | 6 - 7 | 550 | 93 | |
NM_065293.6 | NP_497694.3 | 7 - 8 | 781 | 93 | ||
NM_065294.6 | NP_497695.3 | 7 - 8 | 782 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |