Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267939.3 | NP_001254868.1 | 20 - 21 | 3270 | 86 | |
NM_065293.6 | NP_497694.3 | 21 - 22 | 3501 | 86 | ||
NM_065294.6 | NP_497695.3 | 19 - 20 | 2974 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |