Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065318.4 | NP_497719.1 | 1 - 2 | 90 | 108 | |
GenBank mRNA | AF082012.1 | - | 1 - 2 | 84 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |