Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tir-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381811.1 | NP_001368535.1 | 1 - 2 | 84 | 75 | |
NM_001384059.1 | 3 - 4 | 393 | 75 | |||
NM_065383.4 | NP_497784.1 | 3 - 4 | 410 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |