Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065401.5 | NP_497802.2 | 1 - 2 | 111 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |