Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027413.4 | NP_001022584.1 | 5 - 6 | 519 | 105 | |
NM_001381825.1 | NP_001366708.1 | 5 - 6 | 533 | 105 | ||
GenBank mRNA | U27124.1 | - | 4 - 5 | 315 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |