Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027473.4 | NP_001022644.1 | 10 - 11 | 2013 | 117 | |
NM_001027475.3 | NP_001022646.1 | 10 - 11 | 1784 | 117 | ||
NM_001267984.4 | NP_001254913.1 | 2 - 3 | 260 | 117 | ||
NM_001384070.1 | 9 - 10 | 1734 | 117 | |||
GenBank mRNA | AF031838.1 | - | 9 - 10 | 1744 | 117 | |
AF031839.1 | - | 9 - 10 | 1741 | 117 | ||
AF031840.1 | - | 10 - 11 | 1791 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |