Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hse-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065475.7 | NP_497876.2 | 6 - 7 | 1104 | 94 | |
GenBank mRNA | AY552607.1 | - | 6 - 7 | 1096 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |