Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndg-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027447.6 | NP_001022618.1 | 5 - 6 | 896 | 124 | |
NM_001027448.4 | NP_001022619.1 | 5 - 6 | 896 | 124 | ||
GenBank mRNA | AF173373.1 | - | 5 - 6 | 894 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |