Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atf-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001384076.1 | 2 - 3 | 675 | 56 | ||
NM_001384077.1 | 3 - 4 | 774 | 56 | |||
NM_065512.4 | NP_497913.1 | 3 - 4 | 558 | 56 | ||
NM_065513.5 | NP_497914.2 | 3 - 4 | 558 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |