Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sel-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027391.8 | NP_001022562.1 | 10 - 11 | 1839 | 103 | |
NM_001027392.6 | NP_001022563.1 | 10 - 11 | 1839 | 103 | ||
GenBank mRNA | AF256466.1 | - | 10 - 11 | 1829 | 103 | |
AF256467.1 | - | 10 - 11 | 1829 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |