Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbg-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065578.6 | NP_497979.1 | 3 - 4 | 288 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |