Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coq-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065613.9 | NP_498014.2 | 3 - 4 | 501 | 76 |
Gene Symbol: C35D10.17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381846.1 | NP_001367631.1 | 1 - 2 | 201 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |