Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065621.9 | NP_498022.1 | 3 - 4 | 507 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |