Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065638.6 | NP_498039.1 | 15 - 16 | 4375 | 96 | |
NM_065639.6 | NP_498040.1 | 15 - 16 | 4439 | 96 | ||
NM_065640.6 | NP_498041.1 | 7 - 8 | 2167 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |