Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065638.6 | NP_498039.1 | 9 - 10 | 2497 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |