Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nca-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136363.4 | NP_001129835.1 | 13 - 14 | 2941 | 99 | |
NM_065653.6 | NP_498054.2 | 14 - 15 | 3097 | 99 | ||
GenBank mRNA | AY555272.2 | - | 14 - 15 | 3097 | 99 | |
DQ917241.1 | - | 13 - 14 | 2979 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |