Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nca-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136363.4 | NP_001129835.1 | 15 - 16 | 3447 | 115 | |
NM_065653.6 | NP_498054.2 | 16 - 17 | 3603 | 115 | ||
GenBank mRNA | AY555272.2 | - | 16 - 17 | 3603 | 115 | |
DQ917241.1 | - | 15 - 16 | 3485 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |