Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065668.6 | NP_498069.1 | 6 - 7 | 1564 | 123 | |
GenBank mRNA | AB023497.1 | - | 6 - 7 | 1564 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |