Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gop-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065718.8 | NP_498119.1 | 5 - 6 | 848 | 73 | |
GenBank mRNA | AY052771.1 | - | 5 - 6 | 848 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |