Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hap-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065720.4 | NP_498121.1 | 1 - 2 | 86 | 95 | |
GenBank mRNA | AY052772.1 | - | 1 - 2 | 86 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |