Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: pqe-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047389.4 | NP_001040854.1 | 1 - 2 | 82 | 103 | |
NM_001382869.1 | 1 - 2 | 83 | 103 | |||
NM_065734.8 | NP_498135.2 | 1 - 2 | 89 | 103 | ||
NM_065735.6 | NP_498136.2 | 1 - 2 | 83 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |