Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065802.9 | NP_498203.2 | 3 - 4 | 718 | 85 | |
GenBank mRNA | AF061513.1 | - | 3 - 4 | 721 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |