Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025977.6 | NP_001021148.1 | 6 - 7 | 672 | 80 | |
NM_001025978.3 | NP_001021149.1 | 6 - 7 | 669 | 80 | ||
NM_065810.7 | NP_498211.1 | 6 - 7 | 669 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |