Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025977.6 | NP_001021148.1 | 9 - 10 | 1317 | 89 | |
NM_001322590.3 | NP_001309588.1 | 8 - 9 | 1261 | 89 | ||
NM_001330884.3 | NP_001317770.1 | 4 - 5 | 657 | 89 | ||
NM_065810.7 | NP_498211.1 | 9 - 10 | 1314 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |