Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025981.3 | NP_001021152.1 | 1 - 2 | 285 | 99 | |
NM_001025982.5 | NP_001021153.1 | 4 - 5 | 422 | 99 | ||
GenBank mRNA | AB016838.4 | - | 1 - 2 | 285 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |