Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025981.3 | NP_001021152.1 | 6 - 7 | 1262 | 72 | |
NM_001025982.5 | NP_001021153.1 | 9 - 10 | 1399 | 72 | ||
GenBank mRNA | AB016838.4 | - | 6 - 7 | 1262 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |