Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027436.3 | NP_001022607.1 | 14 - 15 | 4340 | 116 | |
NM_001306735.2 | NP_001293664.1 | 11 - 12 | 4195 | 116 | ||
NM_001306736.3 | NP_001293665.1 | 11 - 12 | 4186 | 116 | ||
NM_001306739.3 | NP_001293668.1 | 15 - 16 | 4504 | 116 | ||
NM_001306740.3 | NP_001293669.1 | 15 - 16 | 4513 | 116 | ||
NM_001306741.3 | NP_001293670.1 | 13 - 14 | 4184 | 116 | ||
NM_001372593.2 | NP_001360234.1 | 10 - 11 | 4013 | 116 | ||
NM_001372595.2 | NP_001360237.1 | 9 - 10 | 3866 | 116 | ||
NM_001382875.1 | 14 - 15 | 4327 | 116 | |||
GenBank mRNA | U25032.1 | - | 14 - 15 | 4339 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |