Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027436.3 | NP_001022607.1 | 3 - 4 | 343 | 83 | |
NM_001306739.3 | NP_001293668.1 | 3 - 4 | 343 | 83 | ||
NM_001306740.3 | NP_001293669.1 | 3 - 4 | 343 | 83 | ||
NM_001306741.3 | NP_001293670.1 | 3 - 4 | 343 | 83 | ||
NM_001379719.1 | NP_001368259.1 | 2 - 3 | 262 | 83 | ||
NM_001382875.1 | 3 - 4 | 339 | 83 | |||
GenBank mRNA | U25032.1 | - | 3 - 4 | 342 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |