Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047399.4 | NP_001040864.1 | 10 - 11 | 1484 | 66 | |
NM_001047400.4 | NP_001040865.1 | 3 - 4 | 354 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |