Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047405.6 | NP_001040870.1 | 4 - 5 | 429 | 123 | |
NM_001379736.1 | NP_001367783.1 | 5 - 6 | 524 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |