Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nfm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027400.4 | NP_001022571.1 | 7 - 8 | 1385 | 71 | |
NM_065934.5 | NP_498335.1 | 7 - 8 | 1811 | 71 | ||
NM_065935.5 | NP_498336.1 | 7 - 8 | 1775 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |