Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065964.1 | NP_498365.1 | 3 - 4 | 849 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |