Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379768.1 | NP_001367476.1 | 2 - 3 | 370 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |