Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025999.4 | NP_001021170.1 | 11 - 12 | 1611 | 74 | |
NM_001026000.4 | NP_001021171.1 | 11 - 12 | 1581 | 74 | ||
NM_001306692.3 | NP_001293621.1 | 9 - 10 | 944 | 74 | ||
NM_001372610.2 | NP_001360187.1 | 9 - 10 | 938 | 74 | ||
NM_001382898.1 | 9 - 10 | 968 | 74 | |||
GenBank mRNA | AY314776.1 | - | 11 - 12 | 1581 | 74 | |
AY314777.1 | - | 11 - 12 | 1611 | 74 | ||
AY314778.1 | - | 11 - 12 | 1611 | 74 | ||
EF141316.1 | - | 8 - 9 | 889 | 74 | ||
EF141317.1 | - | 8 - 9 | 859 | 74 | ||
EF141318.1 | - | 8 - 9 | 889 | 74 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |