Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: odr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382900.1 | 4 - 5 | 878 | 135 | ||
GenBank mRNA | AF055911.1 | - | 3 - 4 | 783 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |