Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066088.7 | NP_498489.2 | 4 - 5 | 699 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |