Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066101.7 | NP_498502.3 | 9 - 10 | 1045 | 85 | |
GenBank mRNA | AY523518.1 | - | 8 - 9 | 927 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |