Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027452.4 | NP_001022623.1 | 2 - 3 | 337 | 57 | |
GenBank mRNA | Z12116.1 | - | 398 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |